Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270046.2 | NP_001256975.1 | 7 - 8 | 1622 | 99 | |
| NM_001374936.2 | NP_001362085.1 | 20 - 21 | 3721 | 99 | ||
| NM_001380893.1 | NP_001367765.1 | 17 - 18 | 3502 | 99 | ||
More Information
Set Membership: |
|

ifc-2
Genomic Map