Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374936.2 | NP_001362085.1 | 4 - 5 | 306 | 96 | |
| NM_001374938.1 | NP_001362002.1 | 5 - 6 | 379 | 96 | ||
| NM_001380893.1 | NP_001367765.1 | 1 - 2 | 87 | 96 | ||
More Information
Set Membership: |
|

ifc-2
Genomic Map