Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374936.2 | NP_001362085.1 | 7 - 8 | 764 | 137 | |
| NM_001374938.1 | NP_001362002.1 | 8 - 9 | 837 | 137 | ||
| NM_001380893.1 | NP_001367765.1 | 4 - 5 | 545 | 137 | ||
More Information
Set Membership: |
|

ifc-2
Genomic Map