Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374936.2 | NP_001362085.1 | 10 - 11 | 1334 | 104 | |
| NM_001374938.1 | NP_001362002.1 | 11 - 12 | 1407 | 104 | ||
| NM_001380893.1 | NP_001367765.1 | 7 - 8 | 1115 | 104 | ||
More Information
Set Membership: |
|

ifc-2
Genomic Map