Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374936.2 | NP_001362085.1 | 12 - 13 | 2091 | 73 | |
| NM_001374938.1 | NP_001362002.1 | 13 - 14 | 2164 | 73 | ||
| NM_001380893.1 | NP_001367765.1 | 9 - 10 | 1872 | 73 | ||
More Information
Set Membership: |
|

ifc-2
Genomic Map