Assay Details
| Gene Symbol: ncam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171615.7 | NP_741707.3 | 12 - 13 | 3857 | 73 | |
| NM_171617.5 | NP_741709.1 | 11 - 12 | 2881 | 73 | ||
| GenBank mRNA | AF456360.2 | - | 12 - 13 | 2765 | 73 | |
More Information
Set Membership: |
|

ncam-1
Genomic Map