Assay Details
| Gene Symbol: ncam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171615.7 | NP_741707.3 | 13 - 14 | 3967 | 154 | |
| NM_171616.5 | NP_741708.1 | 12 - 13 | 2897 | 154 | ||
| NM_171617.5 | NP_741709.1 | 12 - 13 | 2991 | 154 | ||
| GenBank mRNA | AF456360.2 | - | 13 - 14 | 2875 | 154 | |
More Information
Set Membership: |
|

ncam-1
Genomic Map