Assay Details
| Gene Symbol: ncam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171615.7 | NP_741707.3 | 9 - 10 | 2540 | 68 | |
| NM_171616.5 | NP_741708.1 | 9 - 10 | 1424 | 68 | ||
| NM_171617.5 | NP_741709.1 | 8 - 9 | 1564 | 68 | ||
| GenBank mRNA | AF456360.2 | - | 9 - 10 | 1448 | 68 | |
More Information
Set Membership: |
|

ncam-1
Genomic Map