Assay Details
Gene Symbol: zfp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029771.6 | NP_001024942.1 | 2 - 3 | 246 | 64 | |
NM_171627.9 | NP_741719.2 | 2 - 3 | 246 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
