Assay Details
| Gene Symbol: zfp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029771.6 | NP_001024942.1 | 5 - 6 | 754 | 75 | |
| NM_001383527.1 | 5 - 6 | 2305 | 75 | |||
| NM_171627.9 | NP_741719.2 | 5 - 6 | 745 | 75 | ||
| NM_171628.3 | NP_741720.2 | 3 - 4 | 504 | 75 | ||
More Information
Set Membership: |
|

zfp-3
Genomic Map