Assay Details
Gene Symbol: zfp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029771.6 | NP_001024942.1 | 6 - 7 | 1396 | 65 | |
NM_001383527.1 | 6 - 7 | 2947 | 65 | |||
NM_171627.9 | NP_741719.2 | 6 - 7 | 1387 | 65 | ||
NM_171628.3 | NP_741720.2 | 4 - 5 | 1146 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
