Assay Details
| Gene Symbol: zfp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029771.6 | NP_001024942.1 | 7 - 8 | 1567 | 84 | |
| NM_001383527.1 | 7 - 8 | 3118 | 84 | |||
| NM_171627.9 | NP_741719.2 | 7 - 8 | 1558 | 84 | ||
| NM_171628.3 | NP_741720.2 | 5 - 6 | 1317 | 84 | ||
More Information
Set Membership: |
|

zfp-3
Genomic Map