Assay Details
| Gene Symbol: ser-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029202.6 | NP_001024373.1 | 1 - 2 | 332 | 78 | |
| NM_171637.6 | NP_741730.1 | 1 - 2 | 461 | 78 | ||
| NM_171941.7 | NP_741731.1 | 1 - 2 | 375 | 78 | ||
More Information
Set Membership: |
|

ser-7
Genomic Map