Assay Details
| Gene Symbol: unc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348588.4 | NP_001335561.1 | 23 - 24 | 5035 | 98 | |
| NM_001348589.4 | NP_001335563.1 | 23 - 24 | 5035 | 98 | ||
| NM_001348592.5 | NP_001335571.1 | 23 - 24 | 5230 | 98 | ||
| NM_001348593.4 | NP_001335573.1 | 23 - 24 | 5230 | 98 | ||
| NM_001368494.4 | NP_001355464.1 | 29 - 30 | 5596 | 98 | ||
| NM_001368495.4 | NP_001355463.1 | 27 - 28 | 5440 | 98 | ||
| NM_001368496.4 | NP_001355462.1 | 27 - 28 | 5464 | 98 | ||
| NM_001368497.4 | NP_001355494.1 | 23 - 24 | 5363 | 98 | ||
| NM_171638.3 | NP_741732.2 | 23 - 24 | 5220 | 98 | ||
| GenBank mRNA | AY264781.1 | - | 23 - 24 | 5220 | 98 | |
More Information
Set Membership: |
|

unc-2
Genomic Map