Assay Details
| Gene Symbol: unc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348588.4 | NP_001335561.1 | 4 - 5 | 594 | 72 | |
| NM_001348589.4 | NP_001335563.1 | 4 - 5 | 594 | 72 | ||
| NM_001348590.4 | NP_001335560.1 | 4 - 5 | 594 | 72 | ||
| NM_001348591.4 | NP_001335562.1 | 4 - 5 | 594 | 72 | ||
| NM_001348592.5 | NP_001335571.1 | 4 - 5 | 789 | 72 | ||
| NM_001348593.4 | NP_001335573.1 | 4 - 5 | 789 | 72 | ||
| NM_001348594.5 | NP_001335570.1 | 4 - 5 | 789 | 72 | ||
| NM_001348595.4 | NP_001335572.1 | 4 - 5 | 789 | 72 | ||
| NM_001348596.5 | NP_001335516.1 | 2 - 3 | 321 | 72 | ||
| NM_001348597.2 | NP_001335515.1 | 2 - 3 | 321 | 72 | ||
| NM_001368494.4 | NP_001355464.1 | 10 - 11 | 1131 | 72 | ||
| NM_001368495.4 | NP_001355463.1 | 8 - 9 | 975 | 72 | ||
| NM_001368496.4 | NP_001355462.1 | 8 - 9 | 999 | 72 | ||
| NM_001368497.4 | NP_001355494.1 | 4 - 5 | 898 | 72 | ||
| NM_171638.3 | NP_741732.2 | 4 - 5 | 755 | 72 | ||
| GenBank mRNA | AY264781.1 | - | 4 - 5 | 755 | 72 | |
More Information
Set Membership: |
|

unc-2
Genomic Map