Assay Details
| Gene Symbol: unc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348588.4 | NP_001335561.1 | 9 - 10 | 1408 | 71 | |
| NM_001348589.4 | NP_001335563.1 | 9 - 10 | 1408 | 71 | ||
| NM_001348590.4 | NP_001335560.1 | 9 - 10 | 1408 | 71 | ||
| NM_001348591.4 | NP_001335562.1 | 9 - 10 | 1408 | 71 | ||
| NM_001348592.5 | NP_001335571.1 | 9 - 10 | 1603 | 71 | ||
| NM_001348593.4 | NP_001335573.1 | 9 - 10 | 1603 | 71 | ||
| NM_001348594.5 | NP_001335570.1 | 9 - 10 | 1603 | 71 | ||
| NM_001348595.4 | NP_001335572.1 | 9 - 10 | 1603 | 71 | ||
| NM_001348596.5 | NP_001335516.1 | 7 - 8 | 1135 | 71 | ||
| NM_001348597.2 | NP_001335515.1 | 7 - 8 | 1135 | 71 | ||
| NM_001368494.4 | NP_001355464.1 | 15 - 16 | 1969 | 71 | ||
| NM_001368495.4 | NP_001355463.1 | 13 - 14 | 1813 | 71 | ||
| NM_001368496.4 | NP_001355462.1 | 13 - 14 | 1837 | 71 | ||
| NM_001368497.4 | NP_001355494.1 | 9 - 10 | 1736 | 71 | ||
| NM_171638.3 | NP_741732.2 | 9 - 10 | 1593 | 71 | ||
| GenBank mRNA | AY264781.1 | - | 9 - 10 | 1593 | 71 | |
More Information
Set Membership: |
|

unc-2
Genomic Map