Assay Details
| Gene Symbol: klp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373256.3 | NP_001359863.1 | 14 | 2952 | 147 | |
| NM_001373257.3 | NP_001359864.1 | 13 - 14 | 2907 | 147 | ||
More Information
Set Membership: |
|

klp-4
Genomic Map