Assay Details
| Gene Symbol: sax-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029819.7 | NP_001024990.1 | 10 - 11 | 3504 | 76 | |
| GenBank mRNA | AF041053.1 | - | 10 - 11 | 3419 | 76 | |
More Information
Set Membership: |
|

sax-3
Genomic Map