Assay Details
| Gene Symbol: sax-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029819.7 | NP_001024990.1 | 1 - 2 | 158 | 68 | |
| NM_171652.7 | NP_741748.2 | 1 - 2 | 156 | 68 | ||
| GenBank mRNA | AF041053.1 | - | 1 - 2 | 73 | 68 | |
More Information
Set Membership: |
|

sax-3
Genomic Map