Assay Details
| Gene Symbol: dpy-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029694.6 | NP_001024865.1 | 7 - 8 | 916 | 69 | |
| NM_171670.6 | NP_741770.1 | 8 - 9 | 934 | 69 | ||
More Information
Set Membership: |
|

dpy-23
Genomic Map