Assay Details
| Gene Symbol: lgx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373300.2 | NP_001360725.1 | 6 | 1020 | 78 | |
| NM_001373301.2 | NP_001360721.1 | 6 | 1020 | 78 | ||
| NM_001373304.2 | NP_001360724.1 | 7 | 1173 | 78 | ||
| NM_001373305.2 | NP_001360720.1 | 7 | 1173 | 78 | ||
| NM_171690.6 | NP_741796.2 | 7 - 8 | 1228 | 78 | ||
More Information
Set Membership: |
|

lgx-1
Genomic Map