Assay Details
| Gene Symbol: pak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029206.3 | NP_001024377.1 | 10 - 11 | 1569 | 79 | |
| NM_001029207.4 | NP_001024378.1 | 10 - 11 | 1558 | 79 | ||
| NM_001029208.5 | NP_001024379.1 | 9 - 10 | 1412 | 79 | ||
| GenBank mRNA | D83215.1 | - | 10 - 11 | 1555 | 79 | |
More Information
Set Membership: |
|

pak-1
Genomic Map