Assay Details
| Gene Symbol: pak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029206.3 | NP_001024377.1 | 1 - 2 | 93 | 57 | |
| NM_001029207.4 | NP_001024378.1 | 1 - 2 | 91 | 57 | ||
| GenBank mRNA | D83215.1 | - | 1 - 2 | 88 | 57 | |
More Information
Set Membership: |
|

pak-1
Genomic Map