Assay Details
| Gene Symbol: pak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029207.4 | NP_001024378.1 | 4 - 5 | 421 | 71 | |
| NM_001029208.5 | NP_001024379.1 | 3 - 4 | 275 | 71 | ||
| GenBank mRNA | D83215.1 | - | 4 - 5 | 418 | 71 | |
More Information
Set Membership: |
|

pak-1
Genomic Map