Assay Details
| Gene Symbol: pak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029206.3 | NP_001024377.1 | 7 - 8 | 1304 | 99 | |
| NM_001029207.4 | NP_001024378.1 | 7 - 8 | 1293 | 99 | ||
| NM_001029208.5 | NP_001024379.1 | 6 - 7 | 1147 | 99 | ||
| GenBank mRNA | D83215.1 | - | 7 - 8 | 1290 | 99 | |
More Information
Set Membership: |
|

pak-1
Genomic Map