Assay Details
| Gene Symbol: mtp-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129709.3 | NP_001123181.1 | 2 - 3 | 194 | 80 | |
| Gene Symbol: bca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171701.6 | NP_741809.1 | 2 - 3 | 197 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_171701.6 transcripts.
Set Membership: |
|

mtp-18
Genomic Map