Assay Details
| Gene Symbol: cpr-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171708.7 | NP_741818.1 | 3 - 4 | 224 | 91 | |
| GenBank mRNA | L39894.1 | - | 3 - 4 | 221 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpr-6| Gene Symbol: cpr-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171708.7 | NP_741818.1 | 3 - 4 | 224 | 91 | |
| GenBank mRNA | L39894.1 | - | 3 - 4 | 221 | 91 | |
Set Membership: |
|