Assay Details
| Gene Symbol: cpr-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171708.7 | NP_741818.1 | 7 - 8 | 1154 | 112 | |
| GenBank mRNA | L39894.1 | - | 7 - 8 | 1151 | 112 | |
More Information
Set Membership: |
|

cpr-6
Genomic Map