Assay Details
| Gene Symbol: cca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381030.1 | NP_001367691.1 | 17 - 18 | 2441 | 130 | |
| NM_001381031.2 | NP_001367690.1 | 17 - 18 | 2441 | 130 | ||
| NM_171733.5 | NP_741848.1 | 19 - 20 | 2527 | 130 | ||
| GenBank mRNA | AF368920.1 | - | 17 - 18 | 2390 | 130 | |
| AY313898.1 | - | 17 - 18 | 2441 | 130 | ||
| AY313899.1 | - | 17 - 18 | 2441 | 130 | ||
| AY313900.1 | - | 17 - 18 | 2441 | 130 | ||
More Information
Set Membership: |
|

cca-1
Genomic Map