Assay Details
| Gene Symbol: dim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029249.6 | NP_001024420.1 | 7 - 8 | 1090 | 66 | |
| NM_001029250.5 | NP_001024421.1 | 1 - 2 | 136 | 66 | ||
| GenBank mRNA | AF500489.1 | - | 1 - 2 | 143 | 66 | |
| AY095447.1 | - | 7 - 8 | 1109 | 66 | ||
More Information
Set Membership: |
|

dim-1
Genomic Map