Assay Details
| Gene Symbol: kin-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270129.4 | NP_001257058.1 | 7 - 8 | 1190 | 106 | |
| NM_001373393.3 | NP_001359981.1 | 6 - 7 | 1098 | 106 | ||
| NM_001373394.1 | NP_001359980.1 | 4 - 5 | 764 | 106 | ||
More Information
Set Membership: |
|

kin-9
Genomic Map