Assay Details
| Gene Symbol: sma-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375119.3 | NP_001361905.1 | 3 | 650 | 63 | |
| NM_001375120.2 | NP_001361887.1 | 1 - 2 | 569 | 63 | ||
| NM_001375121.3 | NP_001361888.1 | 1 - 2 | 569 | 63 | ||
| NM_001375122.2 | NP_001361889.1 | 1 - 2 | 569 | 63 | ||
| NM_001375123.1 | NP_001361886.1 | 1 - 2 | 569 | 63 | ||
| NM_001375124.1 | NP_001361885.1 | 2 - 3 | 650 | 63 | ||
| NM_001377619.1 | NP_001364627.1 | 2 - 3 | 644 | 63 | ||
| NM_001377620.1 | NP_001364832.1 | 1 - 2 | 569 | 63 | ||
| NM_001383639.1 | 1 - 2 | 569 | 63 | |||
| NM_171771.6 | NP_741896.2 | 1 - 2 | 569 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001375124.1 transcripts.
Set Membership: |
|

sma-9
Genomic Map