Assay Details
| Gene Symbol: sma-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375119.3 | NP_001361905.1 | 5 - 6 | 1752 | 71 | |
| NM_001375120.2 | NP_001361887.1 | 6 - 7 | 2199 | 71 | ||
| NM_001375121.3 | NP_001361888.1 | 6 - 7 | 2199 | 71 | ||
| NM_001375122.2 | NP_001361889.1 | 6 - 7 | 2199 | 71 | ||
| NM_001375123.1 | NP_001361886.1 | 6 - 7 | 2199 | 71 | ||
| NM_001375124.1 | NP_001361885.1 | 7 - 8 | 2280 | 71 | ||
| NM_001375125.2 | NP_001361884.1 | 5 - 6 | 1746 | 71 | ||
| NM_001377619.1 | NP_001364627.1 | 7 - 8 | 1950 | 71 | ||
| NM_001377620.1 | NP_001364832.1 | 6 - 7 | 1875 | 71 | ||
| NM_001383639.1 | 6 - 7 | 1875 | 71 | |||
| NM_171771.6 | NP_741896.2 | 4 - 5 | 1671 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001375125.2 transcripts.
Set Membership: |
|

sma-9
Genomic Map