Assay Details
| Gene Symbol: scd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373531.3 | NP_001360646.1 | 5 | 2156 | 63 | |
| NM_001373535.3 | NP_001360784.1 | 4 - 5 | 1859 | 63 | ||
| NM_001381122.2 | NP_001367017.1 | 3 - 4 | 1754 | 63 | ||
| NM_001381123.2 | NP_001368762.1 | 4 - 5 | 2128 | 63 | ||
| NM_171974.7 | NP_741912.1 | 5 - 6 | 2207 | 63 | ||
More Information
Set Membership: |
|

scd-1
Genomic Map