Assay Details
| Gene Symbol: flp-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171804.7 | NP_741934.1 | 1 - 2 | 173 | 64 | |
| NM_171805.4 | NP_741935.1 | 1 - 2 | 244 | 64 | ||
| GenBank mRNA | AF042394.1 | - | 1 - 2 | 125 | 64 | |
More Information
Set Membership: |
|

flp-8
Genomic Map