Assay Details
| Gene Symbol: pat-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381698.1 | NP_001367550.1 | 8 - 9 | 792 | 70 | |
| NM_001381700.1 | NP_001367551.1 | 11 - 12 | 1101 | 70 | ||
| NM_001381702.1 | NP_001367555.1 | 3 - 4 | 183 | 70 | ||
More Information
Set Membership: |
|

pat-12
Genomic Map