Assay Details
| Gene Symbol: akt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028475.4 | NP_001023646.1 | 5 - 6 | 818 | 89 | |
| GenBank mRNA | AF072380.1 | - | 5 - 6 | 812 | 89 | |
More Information
Set Membership: |
|

akt-1
Genomic Map