Assay Details
| Gene Symbol: flp-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029581.9 | NP_001024752.1 | 2 - 3 | 309 | 61 | |
| GenBank mRNA | AF042397.1 | - | 2 - 3 | 253 | 61 | |
More Information
Set Membership: |
|

flp-11
Genomic Map