Assay Details
| Gene Symbol: ajm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330906.2 | NP_001317734.1 | 16 - 17 | 3189 | 88 | |
| NM_001330908.3 | NP_001317889.1 | 10 - 11 | 2369 | 88 | ||
| NM_001330909.3 | NP_001317890.1 | 10 - 11 | 2144 | 88 | ||
| NM_001330910.3 | NP_001317891.1 | 8 - 9 | 2012 | 88 | ||
| NM_001330911.3 | NP_001317888.1 | 7 - 8 | 1898 | 88 | ||
| NM_001330912.3 | NP_001317897.1 | 6 - 7 | 1582 | 88 | ||
| NM_001330913.3 | NP_001317898.1 | 5 - 6 | 1337 | 88 | ||
| NM_001330914.3 | NP_001317899.1 | 4 - 5 | 1265 | 88 | ||
| NM_001330915.3 | NP_001317736.1 | 2 - 3 | 968 | 88 | ||
| NM_001381045.1 | NP_001368606.1 | 16 - 17 | 3152 | 88 | ||
| NM_001381046.2 | NP_001367383.1 | 16 - 17 | 3160 | 88 | ||
| NM_001381047.2 | NP_001367382.1 | 16 - 17 | 3152 | 88 | ||
| NM_001381048.1 | NP_001367381.1 | 16 - 17 | 3189 | 88 | ||
More Information
Set Membership: |
|

ajm-1
Genomic Map