Assay Details
| Gene Symbol: ttyh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383646.1 | 1 - 2 | 147 | 82 | ||
| NM_077502.7 | NP_509903.1 | 1 - 2 | 148 | 82 | ||
| NM_171973.9 | NP_741904.1 | 1 - 2 | 150 | 82 | ||
More Information
Set Membership: |
|

ttyh-1
Genomic Map