Assay Details
| Gene Symbol: zak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372611.2 | NP_001360014.1 | 7 - 8 | 1349 | 69 | |
| NM_001379778.1 | NP_001367790.1 | 7 - 8 | 1367 | 69 | ||
| NM_001379779.2 | NP_001367792.1 | 7 - 8 | 1372 | 69 | ||
More Information
Set Membership: |
|

zak-1
Genomic Map