Assay Details
| Gene Symbol: tlk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268088.3 | NP_001255017.1 | 5 - 6 | 2554 | 108 | |
| NM_181913.5 | NP_871642.2 | 7 - 8 | 2797 | 108 | ||
More Information
Set Membership: |
|

tlk-1
Genomic Map