Assay Details
| Gene Symbol: tlk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268088.3 | NP_001255017.1 | 3 - 4 | 1016 | 72 | |
| NM_001382967.1 | 5 - 6 | 1377 | 72 | |||
| NM_066744.5 | NP_499145.2 | 6 - 7 | 1447 | 72 | ||
| NM_181913.5 | NP_871642.2 | 5 - 6 | 1259 | 72 | ||
| GenBank mRNA | AY450852.1 | - | 5 - 6 | 1259 | 72 | |
More Information
Set Membership: |
|

tlk-1
Genomic Map