Assay Details
| Gene Symbol: clp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065059.9 | NP_497460.2 | 5 - 6 | 1309 | 73 | |
| NM_181914.6 | NP_871643.1 | 5 - 6 | 1311 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

clp-4| Gene Symbol: clp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065059.9 | NP_497460.2 | 5 - 6 | 1309 | 73 | |
| NM_181914.6 | NP_871643.1 | 5 - 6 | 1311 | 73 | ||
Set Membership: |
|