Assay Details
| Gene Symbol: alh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382859.1 | 3 - 4 | 733 | 64 | ||
| NM_065680.7 | NP_498081.2 | 4 - 5 | 700 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alh-1| Gene Symbol: alh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382859.1 | 3 - 4 | 733 | 64 | ||
| NM_065680.7 | NP_498081.2 | 4 - 5 | 700 | 64 | ||
Set Membership: |
|