Assay Details
| Gene Symbol: cit-1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382929.1 | 5 - 6 | 1725 | 89 | ||
| NM_066344.5 | NP_498745.1 | 6 - 7 | 1520 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cit-1.2| Gene Symbol: cit-1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382929.1 | 5 - 6 | 1725 | 89 | ||
| NM_066344.5 | NP_498745.1 | 6 - 7 | 1520 | 89 | ||
Set Membership: |
|