Assay Details
| Gene Symbol: nono-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372604.2 | NP_001359971.1 | 4 - 5 | 905 | 77 | |
| NM_065894.4 | NP_498295.3 | 4 - 5 | 901 | 77 | ||
| NM_181937.5 | NP_871666.1 | 4 - 5 | 865 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001372604.2 transcripts.
Set Membership: |
|

nono-1
Genomic Map