Assay Details
| Gene Symbol: asd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306717.3 | NP_001293646.1 | 3 - 4 | 486 | 98 | |
| NM_001384065.1 | 4 - 5 | 671 | 98 | |||
| NM_065440.6 | NP_497841.1 | 4 - 5 | 574 | 98 | ||
| GenBank mRNA | DQ485531.1 | - | 4 - 5 | 596 | 98 | |
More Information
Set Membership: |
|

asd-1
Genomic Map