Assay Details
| Gene Symbol: flp-33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381232.1 | NP_001367051.1 | 7 - 8 | 3086 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

flp-33| Gene Symbol: flp-33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381232.1 | NP_001367051.1 | 7 - 8 | 3086 | 79 | |
Set Membership: |
|