Assay Details
| Gene Symbol: cebp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182035.5 | NP_871835.1 | 2 - 3 | 286 | 150 | |
| Gene Symbol: lpd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059405.6 | NP_491806.2 | 2 - 3 | 208 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_059405.6 transcripts.
Set Membership: |
|

cebp-2
Genomic Map